missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.500 Normal (N/2) in Reagent Alcohol, Reagents

Catalog No. 01335085
Change view
Click to view available options
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01335085 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335085 Supplier Labchem Inc Supplier No. CH1036001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hydrochloric Acid, 0.500 Normal (N/2) in Reagent Alcohol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 64-17-5 , 67-63-0 , 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Hydrogen chloride solution
IUPAC Name ethanol
SMILES CCO

Specifications

pH <1
Color Colorless
Boiling Point 78°C
Packaging Glass Bottle
Specific Gravity 0.8163
Quantity 1 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Hydrochloric Acid, 0.500 Normal (N/2) in Reagent Alcohol