missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.1142 N, for Kjeldahl Analysis of Proteins, Ricca Chemical

For Analysis of Proteins (Kjeldahl)

$283.88 - $283.88

Chemical Identifiers

Molecular Formula ClH
Molecular Weight (g/mol) Mixture
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
3610-1
View Documents
Ricca Chemical Company
36101
4 L Natural Poly Bottle
Each for $283.88
Only null left
 
Description

Description

Specifications

Chemical Identifiers

ClH
MFCD00011324 MFCD00792839
313
hydrogen chloride
Mixture
VEXZGXHMUGYJMC-UHFFFAOYSA-N
CHEBI:17883
Cl

Specifications

97.0,1.08
7647-01-0,7732-18-5
4 L
ClH
Natural Poly Bottle
Cl
Mixture
CHEBI:17883
0.1142Normal
Colorless
Liquid
1.13
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
Laboratory
Hydrochloric Acid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'