missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.1142 N, for Kjeldahl Analysis of Proteins, Ricca Tightrant™

Catalog No. 36101
Change view
Click to view available options
Quantity:
4 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
36101 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 36101 missing translation for 'mfr' Ricca Chemical Company missing translation for 'supplierNo' 36101

For Analysis of Proteins (Kjeldahl)

  • Tested in and ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

Molecular Formula ClH
Molecular Weight (g/mol) Mixture
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

CAS Min % 97.0, 1.08
Color Colorless
CAS 7647-01-0, 7732-18-5
Physical Form Liquid
Quantity 4 L
CAS Max % 1.13
Packaging Natural Poly Bottle
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.1142Normal
Chemical Name or Material Hydrochloric Acid