missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.1 N, Aqueous Solution, Spectrum™ Chemical

Catalog No. 18610859
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610859 1 L Poly Bottle
18610860 4 L Poly Bottle
18610861 20 L Poly Bottle
18610862 200 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610859 Supplier Spectrum Chemical Mfg Cor Supplier No. H1201LT
Only null left

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5

Specifications

Concentration 0.100 N ± 0.001 N
CAS Max % 99.64%, 0.36%
Packaging Poly Bottle
Quantity 1 L
Grade Ungraded
Chemical Name or Material Hydrochloric Acid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.