missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.0100 N (N/100), Ricca Chemical

Catalog No. 35904
Change view
Click to view available options
Quantity:
120 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
35904 120 mL Natural Poly Bottle
359032 1 L Natural Poly Bottle
35901 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 35904 Supplier Ricca Chemical Company Supplier No. 35904
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Appearance of Solution Colorless Liquid
CAS Min % 98.0, 0.1
Concentration 0.00995 to 0.01005 N
CAS Max % 100.0, 0.1
Packaging Natural Poly Bottle
Quantity 120 mL
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.0100 N
Assay Normal at 20°C
Chemical Name or Material Hydrochloric Acid
Show More Show Less
ShelfLife 24 months