missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.005 Normal, Reagents

Catalog No. 01334979
Change view
Click to view available options
Quantity:
1 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334979 10 L
01334978 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334979 Supplier Labchem Inc Supplier No. CH16090010F

Hydrochloric Acid, 0.005 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Hydrogen chloride solution
IUPAC Name water
SMILES O

Specifications

pH 2.3
Color Colorless
Boiling Point 100°C
Packaging Cubitainer
Specific Gravity 1.0001
Quantity 10 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Hydrochloric Acid, 0.005 Normal