missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydridotetrakis(triphenylphosphine)rhodium(I)
CAS: 18284-36-1 | C72H61P4Rh | 1153.08 g/mol
$190.77 - $463.37
Chemical Identifiers
| CAS | 18284-36-1 |
|---|---|
| Molecular Formula | C72H61P4Rh |
| Molecular Weight (g/mol) | 1153.08 |
| MDL Number | MFCD00015867 |
| InChI Key | QTTJMGBPWXLZKV-UHFFFAOYSA-N |
| Synonym | hrh pph3 4, hydridotetrakis triphenylphosphine rhodium i, tetrakis triphenylphosphine rhodium hydride, tetrakis triphenylphosphine rhodium, tetrakis triphenylphosphine rhodium i hydride, molecular weight:456.19, molecular formula:c16h26o2rh2, cas number:73468-85-6, rhodium, tetrakis triphenylphosphine, hydridotetrakis triphenylphosphine rhodium |
| PubChem CID | 11981874 |
| IUPAC Name | tetrakis(triphenylphosphane) rhodium |
| SMILES | [RhH].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC377932500
|
Thermo Scientific Chemicals
377932500 |
250 mg | Glass bottle |
Each for $190.77
|
|
||||
|
AC377930010
|
Thermo Scientific Chemicals
377930010 |
1 g | Glass bottle |
Each for $463.37
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
| 18284-36-1 | |
| 1153.08 | |
| QTTJMGBPWXLZKV-UHFFFAOYSA-N | |
| 11981874 | |
| [RhH].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
| C72H61P4Rh | |
| MFCD00015867 | |
| hrh pph3 4, hydridotetrakis triphenylphosphine rhodium i, tetrakis triphenylphosphine rhodium hydride, tetrakis triphenylphosphine rhodium, tetrakis triphenylphosphine rhodium i hydride, molecular weight:456.19, molecular formula:c16h26o2rh2, cas number:73468-85-6, rhodium, tetrakis triphenylphosphine, hydridotetrakis triphenylphosphine rhodium | |
| tetrakis(triphenylphosphane) rhodium |
Specifications
| 18284-36-1 | |
| Ochre | |
| Glass bottle | |
| MFCD00015867 | |
| hrh pph3 4, hydridotetrakis triphenylphosphine rhodium i, tetrakis triphenylphosphine rhodium hydride, tetrakis triphenylphosphine rhodium, tetrakis triphenylphosphine rhodium i hydride, molecular weight:456.19, molecular formula:c16h26o2rh2, cas number:73468-85-6, rhodium, tetrakis triphenylphosphine, hydridotetrakis triphenylphosphine rhodium | |
| [RhH].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 1153.08 | |
| 1151.25 | |
| Hydridotetrakis(triphenylphosphine)rhodium(I) |
| 140°C to 146°C | |
| Authentic | |
| C72H61P4Rh | |
| 250 mg | |
| QTTJMGBPWXLZKV-UHFFFAOYSA-N | |
| tetrakis(triphenylphosphane) rhodium | |
| 11981874 | |
| Crystalline Powder |
RUO – Research Use Only