accessibility menu, dialog, popup

UserName

CAS RN 18284-36-1

IUPAC Name: tetrakis(triphenylphosphane) λ¹-rhodium
Synonyms: hydridotetrakis(triphenylphosphine)rhodium(I)
Molecular Weight (g/mol): 1153.08
Molecular Formula: C72H61P4Rh
InChi Key: QTTJMGBPWXLZKV-UHFFFAOYSA-N
SMILES: [RhH].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results