missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Standard, 100 ppm N2H4, Ricca Chemical

$150.88 - $214.63

Chemical Identifiers

CAS 5341-61-7
Molecular Formula ClH5N2
Molecular Weight (g/mol) 68.50
MDL Number MFCD00064543
InChI Key BIVUUOPIAYRCAP-UHFFFAOYSA-N
PubChem CID 17548
IUPAC Name hydrogen hydrazine chloride
SMILES [H+].[Cl-].NN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
35524
View Documents
Ricca Chemical Company
35524
120 mL Amber Glass Bottle
Each for $150.88
Only null left
 
3552-16
View Documents
Ricca Chemical Company
355216
500 mL Amber Glass Bottle
Each for $214.63
Only null left
 
Description

Description

Specifications

Chemical Identifiers

5341-61-7
68.50
BIVUUOPIAYRCAP-UHFFFAOYSA-N
hydrogen hydrazine chloride
ClH5N2
MFCD00064543
17548
[H+].[Cl-].NN

Specifications

Amber Glass Bottle
97.0,1.14,0.03
1 mL = 0.1 mg N
ClH5N2
Colorless
BIVUUOPIAYRCAP-UHFFFAOYSA-N
hydrogen hydrazine chloride
120 mL
Hydrazine Standard
5341-61-7,7647-01-0,7732-18-5
100.0,1.19,0.03
Liquid
MFCD00064543
100°C
[H+].[Cl-].NN
68.50
17548
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'