missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Solvent, Medium for volumetric two-component Karl Fischer titration (methanol-based), Solstice

Catalog No. 60090267
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60090267 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60090267 Supplier Honeywell Inc Supplier No. 348001LUS
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Methanol-based solvent designed for volumetric two-component Karl Fischer titration.

Advantages of Hydranal two-component reagents:

  • High titration speed
  • Ideal accuracy for small amounts of water
  • High buffer capacity
  • Exact and stable titer
  • E-type reagents: reduced toxicity compared to methanol
  • Long shelf life (three years for titrants, five years for solvents)

Improved Composition

In two-component reagents the Karl Fisher reactants are separated into two solutions: the titrant and the solvent. Hydranal-Titrant contains iodine dissolved in an alcohol with a precisely defined concentration. Hydranal-Solvent is an alcoholic solution of sulfur dioxide and imidazole. The alcohol is either methanol for standard reagents or ethanol for E-type reagents.

Chemical Identifiers

CAS 67-56-1 , 288-32-4 , 7446-09-5
Molecular Formula O2S
Molecular Weight (g/mol) 64.06
InChI Key RAHZWNYVWXNFOC-UHFFFAOYSA-N
IUPAC Name (oxo-λ⁴-sulfanylidene)oxidane
SMILES O=S=O

Specifications

Boiling Point No data available
CAS Min % ≥60.00, ≥10.00
CAS Max % ≤100.00, <30.00
Density 0.890 g/cm3 (20°C)
Physical Form Liquid
Quantity 1 L
Flash Point 11°C
Packaging Polycoated Bottle
Grade Two Component Volumetry
Chemical Name or Material Methanol
Hazard Category Danger
Hazard Statement H370+H360+H332+H314+H225
Precautionary Statement P271+P280+P270+P264+P260+P243+P242+P241+P240+P233+P210+P202+P201
Recommended Storage Room Temperature