Learn More
HYDRANAL™ - Methanol Rapid, Reagent for accelerated volumetric one-component KF titration (working medium), Honeywell Fluka™
Medium for accelerated volumetric one-component Karl Fischer titration.
Supplier:  Honeywell Chemicals 378174X25LUS
 View more versions of this product
Catalog No. 60046664
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
HYDRANAL™ - Methanol Rapid
64°C
<1.0000%,<5.0000%
∼0.80 g/mL
11°C
UN1230
Karl Fischer
Reagent for accelerated volumetric one-component KF titration (working medium)
≥0.1000%,≥1.0000%
288-32-4 , 7446-09-5
4 x 2.5 L
Glass Bottle
Methyl alcohol
Product Suggestions
Product Suggestions