missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-CompoSolver E, Honeywell Fluka™

Medium for volumetric one-component Karl Fischer titration (ethanol-based)

$1041.87 - $1041.87

Specifications

Chemical Name or Material HYDRANAL™-CompoSolver E
CAS 64-17-5 , 693-98-1 , 7446-09-5
Density 0.80g/mL
Flash Point 12°C (53.6°F)
UN Number UN1170
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
60046410
View Documents
Honeywell-Hydranal
347346X1L
6 x 1 L
Case of 6 Each for $1,041.87
Only null left
 
Description

Description

Advantages of Hydranal one-component reagents:

  • Unlimited water capacity
  • Convenient and easy to use
  • The greatest flexibility in working media selection
  • reduced toxicity compared to methanol based systems
  • Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
  • Long shelf life (three years)

Improved Composition

Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.

Specifications

Specifications

HYDRANAL™-CompoSolver E
0.80g/mL
UN1170
64-17-5 , 693-98-1 , 7446-09-5
12°C (53.6°F)
Videos
SDS
Documents

Documents