missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™-Buffer Base, Honeywell Fluka™
Liquid buffer medium for Karl Fischer titration in alkaline samples (based on salicylic acid)
$254.82 - $254.82
Specifications
| Chemical Name or Material | HYDRANAL™-Buffer Base |
|---|---|
| CAS | 69-72-7 , 67-56-1 , 64-17-5 , 121-44-8 |
| Flash Point | 11°C (51.8°F) |
| MDL Number | MFCD00147178 |
| UN Number | UN1993BE |
Description
Advantages of HYDRANAL- Auxiliaries
- Broad product range of solubilizer for special sample application
- Broad product range of buffer for pH related sample application
- Drying agents for cell protection and drying of carrier gasses.