missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Minimal Essential Medium Variations: Powder
Supports cell growth in research and biomanufacturing cell culture processes.
Supplier: Cytiva SH3000802
Description
Specifications
| 2° to 8°C, away from light and moisture | |
| Non-sterile | |
| No Sodium Bicarbonate |
| Powder | |
| Glutamine, None | |
| 2 x 5 L |