missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Hank's 1X Balanced Salt Solutions
Buffer cell culture growth with 1X balanced salt solutions
Supplier: Cytiva SH3003101
Description
- Used for washing and treating tissues and cells
Specifications
| cGMP and ISO 9001:2000 Guidelines | |
| 1 X | |
| Hanks' Balanced Salt Solution | |
| Phenol Red | |
| =1.0EU/mL | |
| 100 mL |
| 15° to 30°C | |
| Liquid | |
| 0.1 μm Sterile-filtered | |
| No Calcium, No Magnesium | |
| 7.0 to 7.4 |