missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Standard
Supplement cell culture growth with U.S. sourced quality cell culture-grade Cytiva HyClone™ Standard Fetal Bovine Serum (FBS)
Supplier: Cytiva SH3008803IR2540
Description
- Support cell culture growth
- Standard grade
- Aseptically packaged in Class 100 cleanroom
Specifications
| Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, IgG | |
| Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
| 500 mL | |
| Sequential-filtered, 100 nm | |
| Irradiated, 25 to 40 doses | |
| No | |
| USDA Inspected |
| Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
| United States | |
| Sterile | |
| Standard | |
| Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
| No |