missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Fetal Bovine Serum (U.S.), Standard

Catalog No. p-8263041
Change view
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Heat Inactivated
Irradiated, 25 to 40 doses
None
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3008802HI 100 mL Heat Inactivated
SH300880340 500 mL Irradiated, 25 to 40 doses
SH3008803 500 mL None
SH3008803HI 500 mL Heat Inactivated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SH3008802HI Supplier Cytiva Supplier No. SH30088.02HI
Only null left

Supplement cell culture growth with U.S. sourced quality cell culture-grade Cytiva HyClone™ Standard Fetal Bovine Serum (FBS)

  • Support cell culture growth
  • Standard grade
  • Aseptically packaged in Class 100 cleanroom

Specifications

Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, IgG
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C
Country of Origin United States
Product Type Fetal Bovine Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Grade Standard
Processing Heat Inactivated
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration For Information Only
Heat Inactivated Yes
Hemoglobin Concentration For Information Only
Mycoplasma Detected No
USDA Safety Testing Included USDA Inspected
Show More Show Less