missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum, Super Low IgG
Enhances cell culture growth, processed via protein G chromatography to substantially reduce immunoglobulin G (IgG) levels
Supplier: Cytiva SH3089802HI
Description
- Processed using protein G chromatography to reduce levels to those required for monoclonal antibody production
- Designed for applications where extremely low levels of inherent bovine IgG are required
- Chromatographically treated to reduce IgG levels to less than 5 μg/mL, while still retaining excellent cell growth properties
- For use in monoclonal antibody production, virus propagation, and immunoassay procedures
Specifications
| <-10°C | |
| Fetal Bovine Serum | |
| Sterile | |
| Heat Inactivated | |
| Monoclonal Antibody Production, Virus Propagation, and Immunoassay Procedures | |
| ≤ 10 mg/dL | |
| HyClone |
| United States | |
| 100 mL | |
| 40 nm filtration | |
| ≤ 10 EU/mL | |
| Yes | |
| ≤ 5 μg/mL |