missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum (New Zealand)

Enhance growth of cells with New Zealand sourced HyClone™ Characterized Fetal Bovine Serum.

$1,106.63

Specifications

Content And Storage Store at or below -10°C
Country of Origin New Zealand
Product Type Fetal Bovine Serum
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Processing Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Processing Price Quantity & Availability  
SH3462IR253
View Documents Promotion Details
Cytiva
SH30406.02IR2535
500 mL Irradiated (25 to 35 doses)
Each for $1,106.63
Only null left
 
Description

Description

  • HyClone Characterized Fetal Bovine Serum (FBS), New Zealand Origin is carefully collected, processed, and filtered in our own New Zealand facility.
  • The great expanse of the Pacific ocean has protected New Zealand’s islands from many outside influences, both geographically and biologically.
  • One of the benefits of this isolation is that New Zealand has the fewest reported bovine diseases in the world, making Characterized FBS, New Zealand Origin an excellent source of bovine serum as it offers greater security.
  • Characterized FBS, New Zealand Origin is filtered through three sequential 100 nm (0.1 μm) pore size-rated filters.
  • Before dispensing, each lot of serum is pooled to ensure uniformity and consistency between bottles (true pool technology).
Specifications

Specifications

Store at or below -10°C
Fetal Bovine Serum
Sequential-filtered, 100 nm
≤ 20 EU/mL
HyClone
New Zealand
Sterile
Characterized
≤ 25 mg/dL
USDA Inspected
Videos
SDS
Documents

Documents