missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Cosmic Calf Serum (U.S. origin)
A bovine calf serum fortified with iron and growth promoting factors for growing a broad spectrum of cell types.
$443.38
Specifications
| Additional Assays | Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG |
|---|---|
| Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C |
| Sterility | Sterile |
| Sterilization Method | Sequential-filtered, 100 nm, Gamma-irradiated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
SH3008704HI
|
Cytiva
SH30087.04HI |
1000 mL |
Each for $443.38
|
|
|||||
Description
HyClone Cosmic Calf serum is a high-performing bovine calf serum that has been fortified with iron and a proprietary blend of growth-promoting factors. The result is a naturally derived, high-quality engineered serum that has demonstrated excellent performance with a broad range of cell types. All components of Cosmic Calf serum are sourced in the USA and are traceable.
Specifications
| Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG | |
| Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C | |
| Sequential-filtered, 100 nm, Gamma-irradiated | |
| Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus |
| Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
| Sterile | |
| Heat Inactivated | |
| Yes |