missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Cosmic Calf Serum (U.S. origin)

Catalog No. SH3008704HI
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3008704HI 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3008704HI Supplier Cytiva Supplier No. SH30087.04HI
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

A bovine calf serum fortified with iron and growth promoting factors for growing a broad spectrum of cell types.

HyClone Cosmic Calf serum is a high-performing bovine calf serum that has been fortified with iron and a proprietary blend of growth-promoting factors. The result is a naturally derived, high-quality engineered serum that has demonstrated excellent performance with a broad range of cell types. All components of Cosmic Calf serum are sourced in the USA and are traceable.

Specifications

Additional Assays Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C
Product Type Calf Bovine Serum
Quantity 1000 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm, Gamma-irradiated
Processing Heat Inactivated
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Heat Inactivated Yes