missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Cell Boost™ 3 Supplement

Fed-batch supplement with amino acids, vitamins, glucose, and trace elements for hybridoma and myeloma cells

$300.15 - $300.15

Specifications

Cell Type Hybridomas, Myelomas
Form Liquid
Product Type Cell Boost Supplement
Format Bottle
For Use With (Application) Improving Productivity and Cell Growth in Fed-Batch Cultures
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
SH3111502
View Documents
Cytiva
SH3111502
1000 mL
Each for $300.15
Only null left
 
Description

Description

  • Chemically Defined (CD), Animal-Derived Component-Free (ADCF), and Protein-Free
  • Supports fed-batch cultures with amino acids, vitamins, glucose, and trace elements
  • Developed for recombinant protein production with various cell lines including hybridoma and myeloma cell lines
  • Designed to support modern cell culture manufacturing processes in research and bioprocessing applications
  • Manufactured to meet cGMP manufacturing standards and QC specifications
  • Available as a ready-to-use liquid feed solution
Specifications

Specifications

Hybridomas, Myelomas
Cell Boost Supplement
Improving Productivity and Cell Growth in Fed-Batch Cultures
Liquid
Bottle
HyClone
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'