missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ CDM4CHO™ Media, Powder
Supplier: Cytiva SH3055601
Specifications
| Powder | |
| Glutamine, Sodium Bicarbonate, Poloxamer 188 | |
| 5 L | |
| 2° to 8°C, away from light and moisture |
| Non-sterile | |
| No Phenol Red, No HEPES, No Protein | |
| cGMP | |
| HyClone CDM4CHO Powder Media - ADCF; 1 x 5L |