missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ CCM1 Cell Culture Media
Protein-containing serum-free media.
Supplier: Cytiva SH3005803
Description
Specifications
| 2° to 8°C, away from light and moisture | |
| Non-sterile | |
| No Sodium Bicarbonate |
| Powder | |
| Phenol Red, Glutamine, HEPES | |
| 10 L |