missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Calf Serum (New Zealand)
$283.42
Specifications
| Additional Assays | Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein |
|---|---|
| Certifications/Compliance | USDA Regulations |
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. |
| Description | Calf Serum (New Zealand) |
| Sterility | Sterile |
Description
Supplement cell culture growth with New Zealand-sourced HyClone™ Bovine Calf Serum, an excellent and cost-effective alternative to fetal bovine serum (FBS).
Specifications
| Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein | |
| Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
| Sterile | |
| ≤ 25 EU/mL | |
| Yes |
| USDA Regulations | |
| Calf Serum (New Zealand) | |
| Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
| Yes |