missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ ActiCHO™ P Powdered Media
Hydrolysate-free and animal-derived component-free (ADCF)
Supplier: Cytiva SH3102508
Description
ActiCHO P powdered cell culture medium is chemically defined, containing poloxamer 188, without insulin or L-glutamine.
- Hydrolysate-free and animal-derived component-free (ADCF). There are no proteins, peptides, or growth factors in ActiCHO P medium.
Specifications
| Powder | |
| Poloxamer 188 | |
| 1 L |
| Non-sterile | |
| No Glutamine, No Insulin | |
| 2 to 8°C, away from light and moisture |