missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ 0.1 N NaOH, Cytiva

Catalog No. SH3088307
Change view
Click to view available options
Quantity:
1 L
10 L
20 L
50 L
100 L
200 L
Packaging:
Single-use Bioprocessing Container
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SH3088307 200 L Single-use Bioprocessing Container
SH3088301 1 L Single-use Bioprocessing Container
SH3088303 10 L Single-use Bioprocessing Container
SH3088304 20 L Single-use Bioprocessing Container
SH3088305 50 L Single-use Bioprocessing Container
SH3088306 100 L Single-use Bioprocessing Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. SH3088307 Supplier Cytiva Supplier No. SH30883.07
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Concentrated sodium hydroxide solutions for bioprocessing applications including pH adjustment and chromatography resin care.

HyClone™ NaOH solutions in single-use bags offer a convenient format to effectively modify pH and properly care for and clean chromatography resins.

  • Manufactured in ISO 9001 and ISO 13485 certified facility.
  • Lot-specific certificate of analysis (CoA) for each lot produced.
  • Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
  • 0.1 μm sterile filtered.
  • All unit sizes are filled in single-use, ready-to-use bags.
  • Raw materials are USP/NF and EP compendial grade.

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

Color Clear, Colorless
Concentration 0.1 M (0.1 N)
Quantity 200 L
Grade NF/USP
Packaging Single-use Bioprocessing Container
Chemical Name or Material Sodium Hydroxide
ShelfLife 12 Months
Recommended Storage 15°C to 30°C