missing translation for 'onlineSavingsMsg'
Learn More

Hyamine Solution, 0.004 M (0.004 N), Ricca Chemical

Catalog No. R3549000500
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Amber Glass Bottle
Safety-coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R3549000500 500 mL Amber Glass Bottle
R354900025D 2.5 L Safety-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R3549000500 Supplier Ricca Chemical Company Supplier No. R3549000500C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121-54-0 , 7732-18-5
Molecular Formula C27H42ClNO2
Molecular Weight (g/mol) 448.088
InChI Key UREZNYTWGJKWBI-UHFFFAOYSA-M
Synonym benzethonium chloride, quatrachlor, hyamine, phemeride, phemerol chloride, benzethoniumchloride, phemithyn, disilyn, kylacol, solamin
PubChem CID 8478
ChEBI CHEBI:31264
IUPAC Name benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride
SMILES CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]

Specifications

Color Colorless
CAS Min % 98.0, 0.18
CAS Max % 100.0, 0.18
Grade Laboratory
Packaging Amber Glass Bottle
pH 5.5 to 7.5
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.004Molar,0.004Normal
Chemical Name or Material Hyamine Solution