missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ HX 531
Description
Potent RXR antagonist (IC50 = 18 nM). Promotes white and brown pre-adipocyte differentiation into white adipocytes. Also inhibits bexarotene -induced brown adipogenic reprogramming of myoblasts.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 483.56 |
| Purity | >98% |
| Molecular Weight (g/mol) | 483.568 |
| Chemical Name or Material | HX 531 |
Safety and Handling
| Recommended Storage | Store at +4°C |