missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human VEGF-C Recombinant Protein, PeproTech®
Recombinant Protein
$125.66 - $4645.76
Specifications
| Accession Number | P49767 |
|---|---|
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 7424 |
| Molecular Weight (g/mol) | 26.2 kDa |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
10020CD100
|
Gibco™
10020CD100UG |
100 μg |
Each for $929.15
|
|
|||||
10020CD1MG
|
Gibco™
10020CD1MG |
1 mg |
Each for $4,645.76
|
|
|||||
10020CD20UG
|
Gibco™
10020CD20UG |
20 μg |
Each for $301.78
|
|
|||||
10020CD250
|
Gibco™
10020CD250UG |
250 μg |
Each for $1,626.02
|
|
|||||
10020CD500
|
Gibco™
10020CD500UG |
500 μg |
Each for $2,789.36
|
|
|||||
10020CD5UG
|
Gibco™
10020CD5UG |
5 μg |
Each for $125.66
|
|
|||||
Description
Recombinant Human VEGF-C is a disulfide-linked homodimeric protein consisting of two 13.5 kDa polypeptide chains of 116 amino acid residues. Due to glycosylation, the protein migrates as a 30.0-33.0 kDa band by SDS-PAGE analysis under non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family, is active in angiogenesis and endothelial cell growth, and can also affect the permeability of blood vessels. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-3 receptors. Only the fully processed form can bind and activate VEGFR-2 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor D.Specifications
| P49767 | |
| protein with no preservative | |
| 26.2 kDa | |
| HEK293 cells | |
| <1 EU/ μg | |
| VEGFC | |
| Determined by its ability to stimulate the proliferation of human microvascular endothelial cells (HMVEC) in low serum conditions. | |
| Recombinant | |
| -20°C | |
| Lyophilized |
| Functional Assay | |
| 7424 | |
| Human VEGF-C | |
| RUO | |
| AW228853; Flt4 ligand; FLT4 ligand DHM; Flt4-L; LMPH1D; Vascular endothelial growth factor C; vascular endothelial growth factor C isoform 129; vascular endothelial growth factor C isoform 184; vascular endothelial growth factor-related protein; VEGF3; VEGFC; VEGF-C; VRP | |
| VEGFC | |
| Unconjugated | |
| AHYNTEILKS IDNEWRKTQC MPREVCIDVG KEFGVATNTF FKPPCVSVYR CGGCCNSEGL QCMNTSTSYL SKTLFEITVP LSQGPKPVTI SFANHTSCRC MSKLDVYRQV HSIIRR | |
| HEK293 cells | |
| ≥ 95% by SDS-PAGE gel and HPLC analyses. |