missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TSLP Uncoated ELISA Kit with Plates

ELISA

$441.00 - $1536.00

Specifications

Accession Number Q969D9
Assay Range 8-1000 pg/mL
Assay Sensitivity 8 pg/mL
Conjugate HRP
Product Type ELISA
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501125208
View Documents
Invitrogen™
88749722
2 x 96 Tests
Each for $441.00
Only null left
 
5017412
View Documents
Invitrogen™
88749786
10 x 96 Tests
Each for $1,110.00
Only null left
 
5017410
View Documents
Invitrogen™
88749776
20 x 96 Tests
Each for $1,536.00
Only null left
 
Description

Description

The Human TSLP Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human TSLP. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

TSLP is a 159 amino acid four helix-bundle hemopoietic cytokine that signals through a heterodimeric receptor complex consisting of the human TSLP receptor and the IL-7R alpha-chain. It impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes, enhances the maturation of CD11c (+) dendritic cells, naive CD4 (+), and CD8 (+) T cells into proallergic effectors, and primes dendritic cells to produce large amounts of IL-12 after CD40 ligand stimulation. TSLP influences the regulation of the positive selection of regulatory T cells and maintenance of peripheral CD4 (+) T cell homeostasis. It induces allergic inflammation by directly activating mast cells and may be involved in the pathophysiology of inflammatory arthritis. It is expressed in heart, liver and prostate.
Specifications

Specifications

Q969D9
8 pg/mL
ELISA
Human
Absorbance Microplate Reader
TSLP
Biotin
Plasma 100 μL, Serum 100 μL, Supernatant 100 μL
thymic stromal derived lymphopoietin
1 hr 45 min
8-1000 pg/mL
HRP
Plasma, Serum, Supernatant
ELISA
85480
Capture Antibody: Pre-titrated, purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86), Purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)
RUO
2-8°C
TSLP
24 hr 30 min
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'