Learn More
Invitrogen™ Human TSLP Uncoated ELISA Kit with Plates
ELISA
Supplier: Invitrogen™ 88749786
Description
The Human TSLP Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human TSLP. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
TSLP is a 159 amino acid four helix-bundle hemopoietic cytokine that signals through a heterodimeric receptor complex consisting of the human TSLP receptor and the IL-7R alpha-chain. It impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes, enhances the maturation of CD11c (+) dendritic cells, naive CD4 (+), and CD8 (+) T cells into proallergic effectors, and primes dendritic cells to produce large amounts of IL-12 after CD40 ligand stimulation. TSLP influences the regulation of the positive selection of regulatory T cells and maintenance of peripheral CD4 (+) T cell homeostasis. It induces allergic inflammation by directly activating mast cells and may be involved in the pathophysiology of inflammatory arthritis. It is expressed in heart, liver and prostate.Specifications
| Q969D9 | |
| 8 pg/mL | |
| HRP | |
| ELISA | |
| Human | |
| Absorbance Microplate Reader | |
| 85480 | |
| Capture Antibody: Pre-titrated, purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86), Purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86) | |
| 10 x 96 Tests | |
| Plasma 100 μL, Serum 100 μL, Supernatant 100 μL | |
| thymic stromal derived lymphopoietin | |
| 1 hr 45 min |
| 8-1000 pg/mL | |
| Capture Antibody: Pre-titrated, purified antibody Detection Antibody: Pre-titrated, biotin-conjugated antibody Standard: Recombinant cytokine for generating standard curve and calibrating samples ELISA Coating Buffer Powder: This uncoated ELISA kit with plates may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 μM)) or 10X PBS ELISA Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20). Assay Diluent: 5X concentrated Detection enzyme: Pre-titrated Avidin-HRP Substrate Solution: Tetramethylbenzidine (TMB) Substrate Solution Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22, -44, -76, -86) |
|
| Uncoated ELISA Kit with Plates | |
| Plasma, Serum, Supernatant | |
| ELISA | |
| Thymic stroma-derived lymphopoietin; thymic stromal lymphopoietin; TSLPThymic stroma-derived lymphopoietin; thymic stromal lymphopoietin; TSLP; BSF-1; IL-4 | |
| TSLP | |
| Biotin | |
| RUO | |
| 2-8°C | |
| TSLP | |
| 24 hr 30 min |