missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TSLP DuoSet ELISA

Catalog No. DY139805
Change view
Click to view available options
Format:
5 96-well Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
DY139805 5 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DY139805 Supplier R&D Systems™ Supplier No. DY139805

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Human TSLP DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human TSLP DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human TSLP.

Specifications

Product Type ELISA Kit
Quantity 1 Kit
Species Human
Conjugate Biotin
Assay Range 31.20 to 2000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Format 5 96-well Plates
For Use With (Application) ELISA
Product Line R&D Systems
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Research Area Cytokines
Show More Show Less