missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TGF-beta 1 DuoSet ELISA

Catalog No. DY24005
Change view
Click to view available options
Packaging Quantity:
5 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DY24005 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DY24005 Supplier R&D Systems™ Supplier No. DY24005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Human TGF-beta 1 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human TGF-beta 1 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human TGF-beta 1.

Specifications

Product Type ELISA Development Kit
Quantity 1 Kit
Species Human
Conjugate Biotin
Assay Range 31.20 to 2000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 7040
Product Line R&D Systems
Synonym CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Packaging Quantity 5 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Research Area Cytokines
Show More Show Less