missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human S9 Fractions

Catalog No. HMS9PL
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HMS9PL 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HMS9PL Supplier Gibco™ Supplier No. HMS9PL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Human S9 Fractions

Pooled human liver S9 fractions.

Total protein concentration: 20 mg/ml

We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.

Related Links:

Specifications

Concentration 20 mg/mL
Product Type Subcellular Fractions
Content And Storage Contains a single 1 mL tube at a concentration of 20 mg/mL. Store between -60°C and -85°C.
Age Adult
Donor Source Pooled
Gender Mixed Gender
Shipping Condition Dry Ice
Sub Cellular Localization S9 Fraction
Target Species Human
Tissue Liver
Form Frozen
Product Line Gibco
Quantity 1 mL
Show More Show Less
Who should I contact if I am looking for hepatocytes with particular characteristics?

Our hepatocytes are prequalified and sold according to application, such as induction, short-term metabolism, and transporter uptake. If you require hepatocytes with particular P450 values or donor specifications, contact our Hepatic Biology Tech Support Team (hepaticproducts@thermofisher.com.com) or (866) 952-3559 to help with specific lot recommendations.

Who can I call if I have technical questions about hepatocyte cell culture?

Thermo Fisher Scientific has a dedicated team to answer questions regarding hepatocyte cell culture. You can send a question by email (hepaticproducts@thermofisher.com) or contact us by phone: US toll-free: (866) 952-3559

Are there guidelines for the procurement of human liver tissue?

Thermofisher Scientific works with a variety of human tissue sources, including Tissue and Organ Procurement Organizations, qualified Research Tissue Organizations and prominent academic and medical centers through collaborations that follow rigorous regulations, certifications and/or accreditations (“Source Facilities”). Tissues obtained through source facilities are consistent with the applicable legal and ethical practices of the United States.

Source Facilities are required by federal law to maintain accurate records and donor confidentiality as well as adhere to certain applicable laws governing tissue used for research globally. These include, but are not limited to:

- 45 CFR Part 46, subparts A, B, C, D where applicable (US federal guidance on the use of human subjects in research)
- Health Insurance Portability and Accountability Act (HIPAA) (addressing issues of confidentiality of personal information)
- National Organ Transplant Act (42 CFR Part 482)
- The Uniform Anatomical Gift Act (UAGA) of 1968, revised 1984

Where do you get your human liver tissue?

The human liver cells are derived from tissue obtained from accredited institutions. Consent is obtained by these institutions from the donor or the donor’s legal next of kin, for the use of the tissue and its derivatives for research purpose.

For Research Use Only. Not for use in diagnostic procedures.