missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | 2900072D10Rik; ECGF; ECGF1; endothelial cell growth factor 1 (platelet-derived); Gliostatin; hPD-ECGF; MEDPS1; MNGIE; MTDPS1; OTTHUMP00000196767; OTTHUMP00000196770; PD TP; PDECGF; PD-ECGF; Pdgfec; platelet derived growth factor, endothelial cell; Platelet-derived endothelial cell growth factor; TdRPase; Thymidine phosphorylase; TP; TYMP |
|---|---|
| Assay Range | 0.4 to 100 ng/mL |
| Assay Sensitivity | 0.4 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH364RBX5
|
Invitrogen™
EH364RBX5 |
5 x 96 Tests |
N/A
|
|
|||||
EH364RB
|
Invitrogen™
EH364RB |
96 Tests |
N/A
|
|
|||||
Description
Human PD-ECGF quantitates human PD-ECGF in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human PD-ECGF.
A platelet-derived endothelial growth factor (PD-ECGF), same as thymidine phosphorylase (TP) or gliostatin. In the presence of inorganic orthophosphate, it catalyses the reversible phospholytic cleavage of thymidine and deoxyuridine to their corresponding bases and 2-deoxyribose-1-phosphate. It is both chemotactic and mitogenic for endothelial cells and a non-heparin binding angiogenic factor present in platelets. It is also involved in transformation of fluoropyrimidines, cytotoxic agents used in the treatment of a variety of malignancies, into active cytotoxic metabolites.Specifications
| 2900072D10Rik; ECGF; ECGF1; endothelial cell growth factor 1 (platelet-derived); Gliostatin; hPD-ECGF; MEDPS1; MNGIE; MTDPS1; OTTHUMP00000196767; OTTHUMP00000196770; PD TP; PDECGF; PD-ECGF; Pdgfec; platelet derived growth factor, endothelial cell; Platelet-derived endothelial cell growth factor; TdRPase; Thymidine phosphorylase; TP; TYMP | |
| 0.4 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| TYMP | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Thymidine Phosphorylase | |
| 4 hrs 45 mins |
| 0.4 to 100 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 1890 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |