missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Osteoadherin/Osteomodulin ELISA Kit
ELISA
$890.00 - $4,420.00
Specifications
| Accession Number | keratan sulfate proteoglycan osteomodulin; KSPG osteomodulin; Omd; OSAD; osteoadherin; osteoadherin proteoglycan; osteomodulin; SLRR2C; UNQ190/PRO216; UNQ190/PRO216SLRR2C |
|---|---|
| Assay Range | 1.4 to 300 ng/mL |
| Assay Sensitivity | 1.4 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH354RBX5
|
Invitrogen™
EH354RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EH354RB
|
Invitrogen™
EH354RB |
96 Tests |
Each for $890.00
|
|
|||||
Description
Human Osteoadherin/Osteomodulin quantitates human Osteoadherin/Osteomodulin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Osteoadherin/Osteomodulin.
OMD may be implicated in biomineralization processes. This protein has a function in binding of osteoblasts via the alpha(V)beta(3)-integrin.Specifications
| keratan sulfate proteoglycan osteomodulin; KSPG osteomodulin; Omd; OSAD; osteoadherin; osteoadherin proteoglycan; osteomodulin; SLRR2C; UNQ190/PRO216; UNQ190/PRO216SLRR2C | |
| 1.4 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| OMD | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| OMD | |
| 4 hrs 45 mins |
| 1.4 to 300 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 4958 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |