missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Osteoadherin/Osteomodulin ELISA Kit
ELISA
Supplier: Invitrogen™ EH354RB
Description
Human Osteoadherin/Osteomodulin quantitates human Osteoadherin/Osteomodulin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Osteoadherin/Osteomodulin.
OMD may be implicated in biomineralization processes. This protein has a function in binding of osteoblasts via the alpha(V)beta(3)-integrin.Chemical Identifiers
| keratan sulfate proteoglycan osteomodulin; KSPG osteomodulin; Omd; OSAD; osteoadherin; osteoadherin proteoglycan; osteomodulin; SLRR2C; UNQ190/PRO216; UNQ190/PRO216SLRR2C | |
| 1.4 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| OMD | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| OMD | |
| 4 hrs 45 mins |
| 1.4 to 300 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 4958 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
Specifications
| keratan sulfate proteoglycan osteomodulin; KSPG osteomodulin; Omd; OSAD; osteoadherin; osteoadherin proteoglycan; osteomodulin; SLRR2C; UNQ190/PRO216; UNQ190/PRO216SLRR2C | |
| 1.4 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| OMD | |
| <10 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
| 1.4 to 300 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 4958 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| OMD | |
| 4 hrs 45 mins |