missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | oligodendrocyte myelin glycoprotein; oligodendrocyte-myelin glycoprotein; Omg; Omgp |
|---|---|
| Assay Range | 0.391 to 25 ng/mL |
| Assay Sensitivity | 0.39 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH353RBX5
|
Invitrogen™
EH353RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EH353RB
|
Invitrogen™
EH353RB |
96 Tests |
Each for $890.00
|
|
|||||
Description
Human Omgp quantitates human Omgp in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Omgp.
Oligodendrocyte myelin glycoprotein (OMG or OMGP) is a cell adhesion molecule contributing to the interactive process required for myelination in the central nervous system. It is also involved in axonogenesis, neuron projection regeneration, and regulation of collateral sprouting of intact axons in response to injury.Specifications
| oligodendrocyte myelin glycoprotein; oligodendrocyte-myelin glycoprotein; Omg; Omgp | |
| 0.39 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| OMG | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| OMG | |
| 4 hrs 45 mins |
| 0.391 to 25 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 4974 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |