missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Myostatin Propeptide Recombinant Protein, PeproTech®
Recombinant Protein
$132.00 - $8480.00
Specifications
| Accession Number | O14793 |
|---|---|
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 2660 |
| Molecular Weight (g/mol) | 27.8 kDa |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
12012100UG
|
Gibco™
12012100UG |
100 μg |
Each for $1,085.28
|
|
|||||
120121MG
|
Gibco™
LS120121MG |
1 mg |
Each for $8,480.00
|
|
|||||
12012250UG
|
Gibco™
12012250UG |
250 μg |
Each for $2,715.00
|
|
|||||
1201225UG
|
Gibco™
LS1201225UG |
25 μg |
Each for $318.00
|
|
|||||
12012500UG
|
Gibco™
12012500UG |
500 μg |
Each for $4,835.00
|
|
|||||
120125UG
|
Gibco™
LS120125UG |
5 μg |
Each for $132.00
|
|
|||||
Description
Recombinant Human Myostatin-Propeptide is a 27.8 kDa protein consisting of 244 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Mature myostatin is obtained by proteolytic processing of a biologically-inactive precursor protein, which contains an N-terminal propeptide of 243 amino acid residues. Myostatin-Propeptide exhibits high binding affinity for myostatin, and has been shown to be a potent inhibitor of myostatin. Over-expression of myostatin-propeptide in mice resulted in large increases (up to 200%) in skeletal muscle mass, similar to those observed in myostatin knockout mice.Specifications
| O14793 | |
| protein with no preservative | |
| 27.8 kDa | |
| E. coli | |
| <1 EU/ μg | |
| pro-Myostatin | |
| Determined by its ability to neutralize the Myostatin inhibitory effect of murine MPC-11 cells. The expected ED50 is 0.01-0.04 ug/ml in the presence of 50 ng/ml Myostatin. | |
| Recombinant | |
| -20°C | |
| Lyophilized |
| Functional Assay | |
| 2660 | |
| Human Myostatin Propeptide | |
| RUO | |
| Myostatin | |
| MSTN | |
| Unconjugated | |
| MNENSEQKEN VEKEGLCNAC TWRQNTKSSR IEAIKIQILS KLRLETAPNI SKDVIRQLLP KAPPLRELID QYDVQRDDSS DGSLEDDDYH ATTETIITMP TESDFLMQVD GKPKCCFFKF SSKIQYNKVV KAQLWIYLRP VETPTTVFVQ ILRLIKPMKD GTRYTGIRSL KLDMNPGTGI WQSIDVKTVL QNWLKQPESN LGIEIKALDE NGHDLAVTFP GPGEDGLNPF LEVKVTDTPK RSRR | |
| E. coli | |
| ≥ 98% by SDS-PAGE gel and HPLC analyses. |