missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | matrix extracellular phosphoglycoprotein; matrix extracellular phosphoglycoprotein with ASARM motif (bone); matrix, extracellular phosphoglycoprotein with ASARM motif (bone); Mepe; OF45; Osteoblast/osteocyte factor 45; Osteoregulin |
|---|---|
| Assay Range | 26 to 2,000 ng/mL |
| Assay Sensitivity | 8.4 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH320RBX5
|
Invitrogen™
EH320RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EH320RB
|
Invitrogen™
EH320RB |
96 Tests |
Each for $890.00
|
|
|||||
Description
Human MEPE/OF45 quantitates human MEPE/OF45 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human MEPE/OF45.
Matrix extracellular phosphoglycoprotein (MEPE), also known as osteoblast/osteocyte factor 45 (OF45), is a member of the small integrin-binding ligand N-linked glycoprotein (SIBLING) family. MEPE promotes renal phosphate excretion and modulates mineralization. It can be phosphorylated by secreted protein kinases. MEPE is expressed in osteoblasts, osteocytes and odontoblasts, and the expression has also been reported in numerous human tumor cell lines. It is a 525-amino acid protein with a calculated molecular weight of 58 kDa.Specifications
| matrix extracellular phosphoglycoprotein; matrix extracellular phosphoglycoprotein with ASARM motif (bone); matrix, extracellular phosphoglycoprotein with ASARM motif (bone); Mepe; OF45; Osteoblast/osteocyte factor 45; Osteoregulin | |
| 8.4 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| MEPE | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| MEPE | |
| 4 hrs 45 mins |
| 26 to 2,000 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 56955 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |