missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-16 (129 aa) Recombinant Protein, PeproTech®
Recombinant Protein
$132.00 - $8480.00
Specifications
| Accession Number | Q14005 |
|---|---|
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 3603 |
| Molecular Weight (g/mol) | 13.3 kDa |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
20016100UG
|
Gibco™
20016100UG |
100 μg |
Each for $1,614.59
|
|
|||||
2001610UG
|
Gibco™
LS2001610UG |
10 μg |
Each for $318.00
|
|
|||||
200161MG
|
Gibco™
LS200161MG |
1 mg |
Each for $8,480.00
|
|
|||||
20016250UG
|
Gibco™
20016250UG |
250 μg |
Each for $2,825.00
|
|
|||||
200162UG
|
Gibco™
LS200162UG |
2 μg |
Each for $132.00
|
|
|||||
20016500UG
|
Gibco™
20016500UG |
500 μg |
Each for $4,835.00
|
|
|||||
2001650UG
|
Gibco™
2001650UG |
50 μg |
Each for $967.23
|
|
|||||
Description
Recombinant Human IL-16 is a 13.3 kDa protein consisting of 129 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The protein encoded by IL-16 is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of IL-16 undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Alternate splicing results in multiple transcript variants.Specifications
| Q14005 | |
| protein with no preservative | |
| 13.3 kDa | |
| E. coli | |
| <1 EU/ μg | |
| IL-16 | |
| Not available | |
| Recombinant | |
| -20°C | |
| Lyophilized |
| ELISA, Functional Assay, Western Blot | |
| 3603 | |
| Human IL-16 (129 aa) | |
| RUO | |
| FLJ16806; FLJ42735; FLJ44234; H-IL-16; il 16; Il16; IL-16; ILN; Interleukin; interleukin 16; Interleukin16; Interleukin-16; LCF; Lymphocyte chemoattractant factor; M-IL-16; mKIAA4048; neuronal interleukin 16; NIL16; PRIL16; prIL-16; prointerleukin 16; Pro-interleukin-16 | |
| IL16 | |
| Unconjugated | |
| PDLNSSTDSA ASASAASDVS VESTAEATVC TVTLEKMSAG LGFSLEGGKG SLHGDKPLTI NRIFKGAASE QSETVQPGDE ILQLGGTAMQ GLTRFEAWNI IKALPDGPVT IVIRRKSLQS KETTAAGDS | |
| E. coli | |
| ≥ 98% by SDS-PAGE gel and HPLC analyses. |