missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-16 (129 aa) Recombinant Protein, PeproTech® DFS Item

Recombinant Protein

Supplier:  Gibco™ 2001650UG

 View more versions of this product

Catalog No. 2001650UG


Only null left
Explore available promotions

Description

Description

Recombinant Human IL-16 is a 13.3 kDa protein consisting of 129 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by IL-16 is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of IL-16 undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Alternate splicing results in multiple transcript variants.
Specifications

Specifications

Q14005
Protein with no preservative
13.3 kDa
Purified
E. coli
RUO
FLJ16806; FLJ42735; FLJ44234; H-IL-16; il 16; Il16; IL-16; ILN; Interleukin; interleukin 16; Interleukin16; Interleukin-16; LCF; Lymphocyte chemoattractant factor; M-IL-16; mKIAA4048; neuronal interleukin 16; NIL16; PRIL16; prIL-16; prointerleukin 16; Pro-interleukin-16
IL16
Protein
Recombinant
-20°C
E. coli
≥ 98% by SDS-PAGE gel and HPLC analyses.
Control, Western Blot Control, Bioactivity
3603
Human IL-16 (129 aa)
50 μg
-20°C
<1 EU/μg
IL-16
Not available
Unconjugated
PDLNSSTDSA ASASAASDVS VESTAEATVC TVTLEKMSAG LGFSLEGGKG SLHGDKPLTI NRIFKGAASE QSETVQPGDE ILQLGGTAMQ GLTRFEAWNI IKALPDGPVT IVIRRKSLQS KETTAAGDS
Not available
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions