missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | CMD1I; CSM1; CSM2; Des; desmin; desmin-like protein; FLJ12025; FLJ39719; FLJ41013; FLJ41793; I79 |
|---|---|
| Assay Range | 2 to 500 ng/mL |
| Assay Sensitivity | 2 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH152RBX5
|
Invitrogen™
EH152RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EH152RB
|
Invitrogen™
EH152RB |
96 Tests |
Each for $890.00
|
|
|||||
Description
Human Desmin quantitates human Desmin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Desmin.
Desmin (approximately 53 kDa) exhibits a high degree of tissue specificity, its expression being predominantly confined to all types of muscle cells (cardiac, skeletal and smooth muscle). Regulation of desmin expression is stage and tissue-specific, since it is induced during terminal development of, for example, skeletal muscle cell differentiation. In skeletal en cardiac muscle cells desmin is localized in the Z-disk region and at the intercalated disk. The expression pattern of desmin in smooth muscle is much more heterogenous. Coexpression of vimentin and desmin has been observed in tumors derived from muscle tissue, i.e. rhabdomyosarcomas and leiomyosarcomas. Furthermore, during myocard dysfunction dramatic changes in the distribution of desmin have been observed.Specifications
| CMD1I; CSM1; CSM2; Des; desmin; desmin-like protein; FLJ12025; FLJ39719; FLJ41013; FLJ41793; I79 | |
| 2 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| DES | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Desmin | |
| 4 hrs 45 mins |
| 2 to 500 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 1674 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |