missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Caspase-1 ELISA Kit DFS Item

ELISA

$880.00 - $4330.00

Specifications

Accession Number CASP1; CASP-1; CASP1 nirs variant 1; caspase 1; caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, convertase); Caspase1; caspase-1; Caspase-1 subunit p10; Caspase-1 subunit p20; ICE; IL-1 beta-converting enzyme; IL-1 B converting enzyme; IL1BC; IL-1 BC; IL1BCE; IL1B-convertase; interleukin 1 beta-converting enzyme; interleukin 1, beta, convertase; interleukin 1-B converting enzyme; Interleukin 1 beta converting enzyme; interleukin-1 beta convertase; interleukin-1 beta-converting enzyme; p45
Assay Range 0.43 to 100 ng/mL
Assay Sensitivity 0.43 ng/mL
Conjugate HRP
Product Type ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
EH70RBX5 DFS Item
missing translation for 'viewDocuments' Promotion Details
Invitrogen™
EH70RBX5
5 x 96 Tests
Each for $4,330.00
Only null left
 
EH70RB DFS Item
missing translation for 'viewDocuments'
Invitrogen™
EH70RB
96 Tests
Each for $880.00
Only null left
 
Description

Description

Human Caspase-1 quantitates human Caspase-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Caspase-1.

Caspases are a family of cysteine proteases that can be divided into the apoptotic and inflammatory caspase subfamilies. Unlike the apoptotic caspases, members of the inflammatory subfamily are generally not involved in cell death but are associated with the immune response to microbial pathogens. Members of this subfamily include caspase-1, -4, -5, and -12 and can activate proinflammatory cytokines such as IL-1-b and IL-18. Caspase-1 was initially identified as an IL-1-b-converting enzyme; later experiments revealed it to be a mammalian homolog of the C. elegans cell death gene ced-3 whose overexpression can induce apoptosis in fibroblasts.
Specifications

Specifications

CASP1; CASP-1; CASP1 nirs variant 1; caspase 1; caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, convertase); Caspase1; caspase-1; Caspase-1 subunit p10; Caspase-1 subunit p20; ICE; IL-1 beta-converting enzyme; IL-1 B converting enzyme; IL1BC; IL-1 BC; IL1BCE; IL1B-convertase; interleukin 1 beta-converting enzyme; interleukin 1, beta, convertase; interleukin 1-B converting enzyme; Interleukin 1 beta converting enzyme; interleukin-1 beta convertase; interleukin-1 beta-converting enzyme; p45
0.43 ng/mL
ELISA
Colorimetric Microplate Reader
CASP1
<10
HRP
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Caspase 1
4 hrs 45 mins
0.43 to 100 ng/mL
HRP
Plasma, Serum, Supernatant
834
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
2°C to 8°C
1 hrs 20 mins
Videos
SDS
Documents

Documents