missing translation for 'onlineSavingsMsg'
Learn More

Genlantis™ Human Bronchial and Tracheal Epithelial Cells and Kits

$2,577.88

Specifications

For Use With (Application) Used to study the regulation of intracellular pH, the stimulant effect of IL-1b on airway epithelial cell growth, the effect of human rhinovirus infection in terms of upregulation of ICAM-1 expression on epithelial cells
Media Recommendation PM511500, PM510500, PM11GS
Quantity >500000 Cells
Source Surface Epithelium of Normal Human Bronchi/Trachea
Sufficient For 16 Population Doublings
View More Specs

Products 2
Catalog Number Mfr. No. Cell Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Cell Type Quantity Price Quantity & Availability  
PH50405A
Encompass
View Documents Promotion Details
Genlantis™
PH50405A
HTEC >500000 Cells
Each for $2,577.88
Only null left
 
PH50205A
Encompass
View Documents Promotion Details
Genlantis™
PH50205A
HBEC >500000 Cells
Each for $2,577.88
Only null left
 
Description

Description

Human Bronchial/Tracheal Epithelial Cells (HBEpC/HTEpC), are derived from the surface epithelium of normal human bronchi. Cells are cryopreserved at first passage and can be cultured and propagated at least 16 population doublings. Airway epithelial cells are targets in asthma, viral infection, and fibrotic lung disease. These cells have been used to study the regulation of intracellular pH, the stimulant effect of IL-1b on airway epithelial cell growth, and the effect of human rhinovirus infection in terms of upregulation of ICAM-1 expression on epithelial cells.

Specifications

Specifications

Used to study the regulation of intracellular pH, the stimulant effect of IL-1b on airway epithelial cell growth, the effect of human rhinovirus infection in terms of upregulation of ICAM-1 expression on epithelial cells
>500000 Cells
16 Population Doublings
PM511500, PM510500, PM11GS
Surface Epithelium of Normal Human Bronchi/Trachea
Videos
SDS
Documents

Documents