missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Genlantis™ Human Bronchial and Tracheal Epithelial Cells and Kits
Description
Human Bronchial/Tracheal Epithelial Cells (HBEpC/HTEpC), are derived from the surface epithelium of normal human bronchi. Cells are cryopreserved at first passage and can be cultured and propagated at least 16 population doublings. Airway epithelial cells are targets in asthma, viral infection, and fibrotic lung disease. These cells have been used to study the regulation of intracellular pH, the stimulant effect of IL-1b on airway epithelial cell growth, and the effect of human rhinovirus infection in terms of upregulation of ICAM-1 expression on epithelial cells.
Specifications
Specifications
| Cell Type | HBEC |
| For Use With (Application) | Used to study the regulation of intracellular pH, the stimulant effect of IL-1b on airway epithelial cell growth, the effect of human rhinovirus infection in terms of upregulation of ICAM-1 expression on epithelial cells |
| Media Recommendation | PM511500, PM510500, PM11GS |
| Quantity | >500000 Cells |
| Source | Surface Epithelium of Normal Human Bronchi/Trachea |
| Sufficient For | 16 Population Doublings |