Learn More
Invitrogen™ Human APRIL ELISA Kit
ELISA
$856.00 - $5960.00
Specifications
| Accession Number | O75888 |
|---|---|
| Assay Range | 0.78 to 50.0 ng/mL |
| Assay Sensitivity | 0.4 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
Includes: Aluminium pouch(es) with a Microwell Plate coated with polyclonal antibody to human APRIL
Biotin-Conjugate anti-human APRIL polyclonal antibody
Streptavidin-HRP
Human APRIL Standard lyophilized, 100ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films
Description
The Human APRIL (Hu A PRoliferation-Inducing Ligand)) ELISA quantitates Hu APRIL in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu APRIL. Principle of the method The Human APRIL solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Members in the TNF superfamily regulate immune responses and induce apoptosis. A novel member in the TNF family was recently identified by several groups and designated APRIL (for A PRoliferation-Inducing Ligand), TALL2 (for TNF and ApoL-related Leukocyte-expressed Ligand 2), and TRDL 1a (for TNF related death ligand 1a) in human and mouse. Two receptors for APRIL were recently identified and designated TACI and BCMA. APRIL stimulates B and T cell proliferation, triggers humoral immune responses, activates NF-kB, and induces cell death. APRIL and its close relative of BlyS and their receptors BCMA and TACI are involved in diseases of autoimmunity and cancer.Specifications
| O75888 | |
| 0.4 ng/mL | |
| ELISA | |
| Human | |
| Colorimetric Microplate Reader | |
| APRIL,CD256,TALL-2,TALL2,TNLG7B,TRDL-1,UNQ383/PRO715,ZTNF2 | |
| 8.1 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| TNF 13, TALL 2, TRDL 1, CD256 antigen | |
| 1 hrs 20 mins |
| 0.78 to 50.0 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| ELISA | |
| 8741 | |
| 7.1% | |
| Pre-coated 96 well plate, Standard, Standard, Assay Buffer concentrate, Assay Buffer concentrate, SAV-HRP, Wash Buffer, Wash Buffer, Chromogen, Chromogen, Stop Solution | |
| RUO | |
| 2°C to 8°C | |
| APRIL | |
| 3 hrs 10 mins |