Learn More
Invitrogen™ Human APRIL ELISA Kit
Description
Includes
Aluminium pouch(es) with a Microwell Plate coated with polyclonal antibody to human APRIL
Biotin-Conjugate anti-human APRIL polyclonal antibody
Streptavidin-HRP
Human APRIL Standard lyophilized, 100ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films
The Human APRIL (Hu A PRoliferation-Inducing Ligand)) ELISA quantitates Hu APRIL in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu APRIL. Principle of the method The Human APRIL solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | O75888 |
| Assay Range | 0.78-50.0 ng/mL |
| Assay Sensitivity | 0.4 ng/mL |
| Content And Storage | Aluminum pouch(es) with a Microplate coated with polyclonal antibody to human APRIL Biotin-Conjugate anti-human APRIL polyclonal antibody Streptavidin-HRP Human APRIL Standard lyophilized, 100 ng/mL upon reconstitution Sample Diluent Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA) Wash Buffer Concentrate 20x (PBS with 1% Tween 20) Substrate Solution (tetramethyl-benzidine) Stop Solution (1M Phosphoric acid) Adhesive Films |
| Conjugate | HRP |
| Detection Method | Colorimetric |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| Show More |