missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ HT ExoSAP-IT™ Fast High-Throughput PCR Product Cleanup
HT ExoSAP-IT Fast High-Throughput PCR Product Clean-up, with its shortened 14 minute protocol, is a highly stable, customized formulation of the original ExoSAP-IT reagent
Supplier: Applied Biosystems™ 785951000RXN
Description
- 14 minute protocol is half the time of standard protocols
- One-tube PCR clean-up - add HT ExoSAP-IT Fast reagent directly to PCR product
- 100% sample recovery - no loss of PCR products regardless of the fragment size
- Stable at 4°C for one month
- Removes excess primers and dNTPs - does not interfere with downstream applications sequencing, SNP analysis, single base extension, fragment analysis
- It is ideal for high-throughput PCR product clean-up using liquid handling platforms and multi-channel pipettes
- Multiple packaging formats (vials, 8-tube strips, 96-well plates) ensure ease of use and flexibility in experimental design
Chemical Identifiers
| -20°C | |
| Fast High-Throughput PCR Product Clean-up | |
| HT ExoSAP-IT™ Fast High-Throughput PCR Product Clean-up | |
| 14 min |
| 8 Tube Strip, 96-well Plate, Vial | |
| For high-throughput PCR product clean-up using liquid handling platforms and multi-channel pipettes | |
| Each |
Specifications
| -20°C | |
| Fast High-Throughput PCR Product Clean-up | |
| 1000 reactions | |
| Each |
| 8 Tube Strip, 96-well Plate, Vial | |
| For high-throughput PCR product clean-up using liquid handling platforms and multi-channel pipettes | |
| HT ExoSAP-IT™ Fast High-Throughput PCR Product Clean-up | |
| 14 min |
For Research Use Only. Not for use in diagnostic procedures.